missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tungstate dihydrate, 99+%, ACS reagent, ACROS Organics™

$51.22 - $424.62

Chemical Identifiers

CAS 10213-10-2
Assay Percent Range 99+%
Molecular Formula Na2O42H2O
Linear Formula Na2WO4·2H2O
Formula Weight 329.85
MDL Number MFCD00149190
Chemical Name or Material Sodium tungstate dihydrate
Grade ACS Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC424471000 SDS Thermo Fisher Scientific
AC424471000
Glass bottle 100g Each for $137.17
AC424470050 SDS Thermo Fisher Scientific
424470050
Glass bottle 5g Each for $51.22
AC424475000 SDS Thermo Fisher Scientific
424475000
Glass bottle 500g Each for $424.62
Specifications

Description

Oxidation reagent
Description & Specifications

Chemical Identifiers

10213-10-2
Na2O42H2O
329.85
Sodium tungstate dihydrate
99+%
Na2WO4·2H2O
MFCD00149190
ACS Reagent Grade

Specifications

Slightly Crystalline Powder or Flakes
100g
15,295
0.001% max.
99.0 to 101.0%
99+%
100ppm max.
10ppm max.
Glass bottle
50ppm max.
ACS Reagent Grade
329.85
White to slightly yellow
15, 8826
0.01% max.
MFCD00149190
Na2O42H2O
0.02 meq/g max.
Na2WO4·2H2O
10213-10-2
Sodium tungstate dihydrate

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning