Please login to your online account to display your discounted pricing

Sodium Tungstate Dihydrate, 99-101%, (ACS Reagent Grade), MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  0215258190

 View more versions of this product

Catalog No. ICN15258190

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Chemical Name or Material Sodium Tungstate Dihydrate
CAS 10213-10-2
Purity Grade ACS Reagent
Assay Percent Range 99-101%
Molecular Formula Na2WO4 · 2H2O
Formula Weight 329.9g/mol
Synonym (Tungstic acid sodium salt)
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Sodium Tungstate Dihydrate
CAS 10213-10-2
Purity Grade ACS Reagent
Assay Percent Range 99-101%
Molecular Formula Na2WO4 · 2H2O
Formula Weight 329.9g/mol
Synonym (Tungstic acid sodium salt)
Accessories

Room Temperature