Please login to your online account to display your discounted pricing

Sodium Tungstate Dihydrate, 99-101%, (ACS Reagent Grade), MP Biomedicals

$430.00 - $430.00

Chemical Identifiers

Chemical Name or Material Sodium Tungstate Dihydrate
CAS 10213-10-2
Purity Grade ACS Reagent
Assay Percent Range 99-101%
Molecular Formula Na2WO4 · 2H2O
Formula Weight 329.9g/mol
Synonym (Tungstic acid sodium salt)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN15258190

 
mp biomedicals
0215258190
500g Each for $430.00
Description & Specifications

Specifications

Quantity 500g
Chemical Name or Material Sodium Tungstate Dihydrate
CAS 10213-10-2
Purity Grade ACS Reagent
Assay Percent Range 99-101%
Molecular Formula Na2WO4 · 2H2O
Formula Weight 329.9g/mol
Synonym (Tungstic acid sodium salt)
Recommended Storage Room Temperature