missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tripolyphosphate, 85%, pure, Tech., ACROS Organics™

Manufacturer:  Fisher Scientific Acros 218670250

 View more versions of this product

Catalog No. AC218670250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Sodium tripolyphosphate
7758-29-4
Crystalline Powder
85%
Na5P3O10
MFCD00003514
STPP
White
tech., 85%
Extra Pure
≥80%
Na5O10P3
367.85
15, 8824
Plastic bottle
25g

Chemical Identifiers

7758-29-4
Na5O10P3
367.85
STPP
≥80%
85%
Na5P3O10
MFCD00003514
Sodium tripolyphosphate, tech., 85%
Extra Pure

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 231-838-7

RTECSNumber : YK4570000

TSCA : TSCA