missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium trimethylsilanolate, 1M solution in THF, AcroSeal™, ACROS Organics™
Chemical Identifiers
CAS | 18027-10-6 |
---|---|
Molecular Formula | C3H9NaOSi |
Molecular Weight (g/mol) | 112.18 |
MDL Number | MFCD00013926 |
InChI Key | HSNUIYJWTSJUMS-UHFFFAOYSA-N |
Synonym | naosi me 3, naotms, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol, trimethyl-, sodium salt, silanol,1,1,1-trimethyl-, sodium salt 1:1, sodiooxytrimethylsilane, sodium trimethylsilanolat, sodium trimethylsilanolate, trimethylsilanol sodium salt, trimethylsilyloxy sodium |
PubChem CID | 23672319 |
IUPAC Name | sodium;trimethyl(oxido)silane |
SMILES | C[Si](C)(C)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC428831000
|
Thermo Fisher Scientific
AC428831000 |
100mL | AcroSeal™ Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC428838000
|
Thermo Fisher Scientific
AC428838000 |
800mL | AcroSeal™ Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
18027-10-6 | |
112.18 | |
HSNUIYJWTSJUMS-UHFFFAOYSA-N | |
23672319 | |
C[Si](C)(C)[O-].[Na+] |
C3H9NaOSi | |
MFCD00013926 | |
naosi me 3, naotms, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol, trimethyl-, sodium salt, silanol,1,1,1-trimethyl-, sodium salt 1:1, sodiooxytrimethylsilane, sodium trimethylsilanolat, sodium trimethylsilanolate, trimethylsilanol sodium salt, trimethylsilyloxy sodium | |
sodium;trimethyl(oxido)silane |
Specifications
18027-10-6,109-99-9 | |
MFCD00013926 | |
HSNUIYJWTSJUMS-UHFFFAOYSA-N | |
sodium;trimethyl(oxido)silane | |
23672319 | |
AcroSeal™ Glass Bottle | |
−17°C |
C3H9NaOSi | |
naosi me 3, naotms, silanol, 1,1,1-trimethyl-, sodium salt 1:1, silanol, trimethyl-, sodium salt, silanol,1,1,1-trimethyl-, sodium salt 1:1, sodiooxytrimethylsilane, sodium trimethylsilanolat, sodium trimethylsilanolate, trimethylsilanol sodium salt, trimethylsilyloxy sodium | |
C[Si](C)(C)[O-].[Na+] | |
112.18 | |
112.18 | |
100mL | |
Sodium trimethylsilanolate |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 241-939-8