Please login to your online account to display your discounted pricing

Sodium trimethylsilanolate, 1M solution in THF, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass bottle

Manufacturer: thermo fisher scientific  AC428838000

 View more versions of this product

Catalog No. AC428838000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 112.18
Quantity 800mL
Chemical Name or Material Sodium trimethylsilanolate
Packaging AcroSeal™ Glass bottle
Molecular Formula C3H9NaOSi
MDL Number MFCD00013926
CAS 18027-10-6
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides.
CAS 109-99-9
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Sodium trimethylsilanolate
CAS 18027-10-6
CAS 109-99-9
Molecular Formula C3H9NaOSi
Formula Weight 112.18
MDL Number MFCD00013926
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. Highly flammable liquid and vapor. May form explosive peroxides.