Please login to your online account to display your discounted pricing

Sodium trimethylsilanolate, 1M solution in dichloromethane, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC428848000

 View more versions of this product

Catalog No. AC428848000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 112.18
Quantity 800mL
Packaging AcroSeal™ Glass Bottle
Density 1.2800g/mL
Chemical Name or Material Sodium trimethylsilanolate
Molecular Formula C3H9NaOSi
MDL Number MFCD00013926
CAS 18027-10-6
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. May cause drowsiness or dizziness.
CAS 75-09-2
Health Hazard 3 GHS P Statement Do not handle until all safety precautions have been read and understood. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Specifications

Chemical Name or Material Sodium trimethylsilanolate
CAS 18027-10-6
CAS 75-09-2
Molecular Formula C3H9NaOSi
Formula Weight 112.18
MDL Number MFCD00013926
Accessories

Danger

GHS P Statement Do not handle until all safety precautions have been read and understood. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

GHS P Statement May cause damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. May cause drowsiness or dizziness.