Learn More
Sodium trifluoromethanesulfonate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02083704
Description
Catalyzes the aza Diels-Alder reaction of Danishefsky's diene with imines in water under neutral conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium trifluoromethanesulfonate | |
253°C to 255°C | |
Odorless | |
CF3SO3Na | |
2 g | |
Hygroscopic | |
Soluble in water (1000g/L). | |
C(F)(F)(F)S(=O)(=O)[O-].[Na+] | |
172.053 | |
172.06 |
2926-30-9 | |
White | |
CF3NaO3S | |
MFCD00061607 | |
3728797 | |
sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d | |
XGPOMXSYOKFBHS-UHFFFAOYSA-M | |
sodium;trifluoromethanesulfonate | |
23676748 | |
Powder |
Chemical Identifiers
2926-30-9 | |
172.053 | |
XGPOMXSYOKFBHS-UHFFFAOYSA-M | |
23676748 | |
C(F)(F)(F)S(=O)(=O)[O-].[Na+] |
CF3NaO3S | |
MFCD00061607 | |
sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d | |
sodium;trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only