missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium trifluoromethanesulfonate

Manufacturer:  Alfa Aesar™ 2083718

 View more versions of this product

Catalog No. AA2083718



Specifications

Specifications

Sodium trifluoromethanesulfonate
Hygroscopic
253°C to 255°C
2926-30-9
CF3SO3Na
3728797
XGPOMXSYOKFBHS-UHFFFAOYSA-M
sodium;trifluoromethanesulfonate
23676748
Powder
Odorless
White
50g
CF3NaO3S
MFCD00061607
sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d
C(F)(F)(F)S(=O)(=O)[O-].[Na+]
172.053
172.06

Chemical Identifiers

2926-30-9
172.053
XGPOMXSYOKFBHS-UHFFFAOYSA-M
23676748
C(F)(F)(F)S(=O)(=O)[O-].[Na+]
CF3NaO3S
MFCD00061607
sodium trifluoromethanesulfonate, sodium triflate, trifluoromethanesulfonic acid sodium salt, sodium trifluoro methane sulfonate, methanesulfonic acid, trifluoro-, sodium salt, pubchem23598, cf3so3na, acmc-1cfau, sodiumtrifluoromethanesulfonate, ksc491a1d
sodium;trifluoromethanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents