missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Trifluoromethanesulfinate, TCI America™

Manufacturer:  TCI America T203325G

 View more versions of this product

Catalog No. T203325G



Specifications

Specifications

Sodium Trifluoromethanesulfinate
25g
CF3NaO2S
sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
C(F)(F)(F)S(=O)[O-].[Na+]
156.054
156.05
≥95.0% (T)
White-Yellow
2926-29-6
MFCD03092989
KAVUKAXLXGRUCD-UHFFFAOYSA-M
sodium;trifluoromethanesulfinate
23690734
Crystal-Powder at 20°C

Chemical Identifiers

2926-29-6
156.054
KAVUKAXLXGRUCD-UHFFFAOYSA-M
23690734
C(F)(F)(F)S(=O)[O-].[Na+]
CF3NaO2S
MFCD03092989
sodium trifluoromethanesulfinate, sodium trifluoromethanesulphinate, sodium trifluoro-methanesulfinate, trifluoromethanesulfinic acid sodium salt, methanesulfinic acid, trifluoro-, sodium salt, langlois reagent, chf3o2s.na, acmc-209h7v, ksc563a2r, sodium trifluoromethylsulfinate
sodium;trifluoromethanesulfinate
Safety and Handling

Safety and Handling

TSCA : No

Documents