missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium trifluoroacetate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1461336

 View more versions of this product

Catalog No. AAA1461336



Description

Description

Applications
Sodium trifluoroacetate is used in acid catalyzed reactions. It is also used as an intermediate for active pharmaceutical ingredients (API). Further, it acts as a precursor for the trifluoromethylation of aldehydes using copper(I) halide as a catalyst.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases. Hygroscopic. Moisture sensitive.
Specifications

Specifications

Sodium trifluoroacetate
500g
98%
MFCD00013217
sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
C(=O)(C(F)(F)F)[O-].[Na+]
136.005
136
~207°C (decomposition)
2923-18-4
C2F3NaO2
3597949
UYCAUPASBSROMS-UHFFFAOYSA-M
sodium;2,2,2-trifluoroacetate
517019
98%

Chemical Identifiers

2923-18-4
136.005
UYCAUPASBSROMS-UHFFFAOYSA-M
517019
C(=O)(C(F)(F)F)[O-].[Na+]
C2F3NaO2
MFCD00013217
sodium trifluoroacetate, trifluoroacetic acid sodium salt, acetic acid, trifluoro-, sodium salt, sodium perfluoroacetate, trifluoroacetate sodium, sodium 2,2,2-trifluoroacetate, trifluoroacetic acid sodium, trifluoroacetic acid,sodium salt, acetic acid, 2,2,2-trifluoro-, sodium salt 1:1, trifluoroacetic acid, sodium salt
sodium;2,2,2-trifluoroacetate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AK0250000

TSCA : TSCA

storageNote1 : Hygroscopic

Documents