missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Triacetoxyborohydride, TCI America™

Manufacturer:  TCI America S039425G

 View more versions of this product

Catalog No. S039425G



Specifications

Specifications

Sodium Triacetoxyborohydride
106°C
56553-60-7
MFCD00012211
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
Crystal-Powder at 20°C
White
25g
C6H11BO6
2813
211.94
≥80.0% (T)

Chemical Identifiers

56553-60-7
C6H11BO6
MFCD00012211
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-4147

TSCA : Yes

Documents