Learn More
Sodium triacetoxyborohydride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 291821000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Sodium Triacetoxyborohydride | |
114.0°C to 118.0°C | |
Authentic | |
Glass Bottle | |
NaB(O2CCH3)3H | |
100 g | |
12,453; 13,283; 06,553; 16,309 | |
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate | |
HHYFEYBWNZJVFQ-UHFFFAOYSA-N | |
sodium bis(acetyloxy)boranuidyl acetate | |
211.94 | |
Powder |
56553-60-7 | |
White | |
96% min | |
C6H10BNaO6 | |
MFCD00012211 | |
3396 | |
15, 8822 | |
Solubility in water: reacts. Other solubilities: soluble in thf,acetonitrile,1,2-dichloroethane | |
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O | |
211.94 | |
97% |
Chemical Identifiers
56553-60-7 | |
211.94 | |
HHYFEYBWNZJVFQ-UHFFFAOYSA-N | |
sodium bis(acetyloxy)boranuidyl acetate |
C6H10BNaO6 | |
MFCD00012211 | |
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate | |
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
RUO – Research Use Only