missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium triacetoxyborohydride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2206022

 View more versions of this product

Catalog No. AAB2206022



Description

Description

Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.

Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
Specifications

Specifications

Sodium triacetoxyborohydride
∼110°C (decomposition)
MFCD00012211
UN3396
Moisture sensitive
Soluble in dimethyl sulfoxide,methanol,benzene,toluene,terahydrofuran,dioxane and methylene chloride.
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O
211.94
95%
56553-60-7
C6H10BNaO6
100 g
4047608
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
HHYFEYBWNZJVFQ-UHFFFAOYSA-N
sodium bis(acetyloxy)boranuidyl acetate
211.94

Chemical Identifiers

56553-60-7
211.94
HHYFEYBWNZJVFQ-UHFFFAOYSA-N
sodium bis(acetyloxy)boranuidyl acetate
C6H10BNaO6
MFCD00012211
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O
Safety and Handling

Safety and Handling

GHS H Statement
H261-H228-H318-H335-H315
In contact with water releases flammable gas.
Flammable solid.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P223-P231+P232-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P335+P334-P362-P370+P378q-P501c

H260-H315-H318

DOTInformation : Hazard Class: 4.3; Packaging Group: II

EINECSNumber : 000-000-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only