Please login to your online account to display your discounted pricing

Sodium tri-sec-butylborohydride, 1M solution in tetrahydrofuran, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Sodium tri-sec-butylborohydride
CAS 67276-04-4
CAS 109-99-9
Molecular Formula C12H28BNa
Linear Formula NaB[CH(CH3)C2H5]3H
Formula Weight 206.15
MDL Number MFCD00011709
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC200025000

  SDS
thermo fisher scientific
AC200025000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.15
Physical Form Liquid
Quantity 500mL
Color Clear colorless to yellow
Density 0.8900g/mL
Linear Formula NaB[CH(CH3)C2H5]3H
CAS 109-99-9
Chemical Name or Material Sodium tri-sec-butylborohydride
CAS 67276-04-4
Health Hazard 3 GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co
Packaging Glass bottle
Molecular Formula C12H28BNa
MDL Number MFCD00011709
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. May form explosive peroxides. Reacts violently

Reduction reagent

Safety and Handling

Danger

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement May cause respiratory irritation. Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. May form explosive peroxides. Reacts violently