Learn More
Sodium tin(IV) oxide hydrate, Reagent Grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01402136
Description
Sodium tin(IV) oxide is used as a mordant, fire retardant textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium tin(IV) oxide hydrate | |
12058-66-1 | |
500 g | |
MFCD00014241 | |
sodium stannate, sodium tin oxide, tin sodium oxide, sodium stannate iv, disodium tin trioxide, unii-jc8111zn4w, stannate sno32-, disodium, disodium stannate, disodium dioxido oxo tin, natriumstannat | |
TVQLLNFANZSCGY-UHFFFAOYSA-N | |
disodium oxostannanebis(olate) | |
25501 | |
Reagent | |
Air Sensitive |
140°C (decomposition) | |
Crystalline | |
Na2O3Sn | |
14,8677 | |
Slightly soluble in water. | |
[Na+].[Na+].[O-][Sn]([O-])=O | |
212.69 | |
212.67 (Anhydrous) | |
Odorless |
Chemical Identifiers
12058-66-1 | |
212.69 | |
TVQLLNFANZSCGY-UHFFFAOYSA-N | |
25501 |
Na2O3Sn | |
MFCD00014241 | |
sodium stannate, sodium tin oxide, tin sodium oxide, sodium stannate iv, disodium tin trioxide, unii-jc8111zn4w, stannate sno32-, disodium, disodium stannate, disodium dioxido oxo tin, natriumstannat | |
[Na+].[Na+].[O-][Sn]([O-])=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 235-030-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only