Please login to your online account to display your discounted pricing

Sodium Thiosulfate Pentahydrate Solution (1N/Certified), Fisher Chemical


Standardized at 25°C against NIST™ potassium dichromate

Quantity 4L
Packaging Poly Bottle

Manufacturer: fisher scientific company  SS3644

 View more versions of this product

Catalog No. SS364-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 4L
Physical Form Liquid
Viscosity [NULL]
Vapor Pressure 14mmHg
NIST Traceable to NIST.
Color Colorless
ChemAlert Storage Symbol Gray
Health Hazard 1 Signal Word:CAUTION!
Health Hazard 2 CAUTION!
Identification Pass Test
CAS 7732-18-5
Traceability to NIST 136F POTASSIUM DICHROMATE
MDL Number MFCD00003499
Packaging Poly Bottle
Chemical Name or Material Sodium Thiosulfate Pentahydrate Solution
Concentration 0.995 to 1.005N
CAS 10102-17-7
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0
Name Note 1N

Specifications

Chemical Name or Material Sodium Thiosulfate Pentahydrate Solution
Name Note 1N
CAS 10102-17-7
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00003499
Accessories

Accessories for Catalog Number: SS3644

Item Mfr. No. Item Description Price Per Unit Quantity  
Accessories
 

Signal Word:CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0

CAUTION!

SDS & Safety Documents