Please login to your online account to display your discounted pricing

Sodium Thiosulfate Pentahydrate Solution (1N/Certified), Fisher Chemical

Standardized at 25°C against NIST™ potassium dichromate

$132.85 - $622.15

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate Pentahydrate Solution
Name Note 1N
CAS 10102-17-7
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00003499
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SS364-1

  SDS
fisher scientific company
FLSS364-1
Poly Bottle 1L Each for $132.85
Case of 6 Each for $797.10

SS364-20

  SDS
fisher scientific company
SS36420
PolyPac™ 20L Each for $622.15

SS364-4

  Accessories SDS
fisher scientific company
SS3644
Poly Bottle 4L Each for $234.08
Case of 4 Each for $936.32
Description & Specifications

Specifications

Chemical Name or Material Sodium Thiosulfate Pentahydrate Solution
Name Note 1N
CAS 10102-17-7
ChemAlert Storage Symbol Gray
Health Hazard 1 Signal Word:CAUTION!
Health Hazard 2 CAUTION!
CAS 7732-18-5
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0
Purity Grade Certified Grade
Physical Form Liquid
Concentration 0.995 to 1.005N
MDL Number MFCD00003499
Identification Pass Test
Packaging Poly Bottle
Traceability to NIST 136F POTASSIUM DICHROMATE
Vapor Pressure 14mmHg
NIST Traceable to NIST.
Boiling Point 100°C
Color Colorless
Melting Point 0°C
Quantity 1L

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Safety and Handling
 

Signal Word:CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0

CAUTION!