Please login to your online account to display your discounted pricing

Sodium Thiosulfate Pentahydrate Solution (0.2N), Fisher Chemical


Standardized at 25°C against NIST™ potassium dichromate

Quantity 20L
Packaging PolyPac™

Manufacturer: fisher scientific company  SS81720

 View more versions of this product

Catalog No. SS817-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Melting Point 0°C
Quantity 20L
Physical Form Liquid
NIST Traceable to NIST.
Vapor Pressure 14mmHg
Color Colorless
Identification Pass Test
CAS 7732-18-5
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0
ChemAlert Storage Symbol Gray
Health Hazard 1 Signal Word:CAUTION!
MDL Number MFCD00003499
Packaging PolyPac™
CAS 10102-17-7
Traceability to NIST LOT NUMBER USED POTASSIUM DICHROMATE
Health Hazard 2 CAUTION!
Concentration 0.1995 to 0.2005
Name Note 0.2N
Chemical Name or Material Sodium Thiosulfate Solution

Specifications

Chemical Name or Material Sodium Thiosulfate Solution
Name Note 0.2N
CAS 10102-17-7
CAS 7732-18-5
MDL Number MFCD00003499
Accessories
 

Signal Word:CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0

CAUTION!

SDS & Safety Documents