Please login to your online account to display your discounted pricing

Sodium Thiosulfate Pentahydrate Solution (0.2N), Fisher Chemical

Standardized at 25°C against NIST™ potassium dichromate

$101.77 - $540.00

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate Solution
Name Note 0.2N
CAS 10102-17-7
CAS 7732-18-5
MDL Number MFCD00003499
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SS817-4

  SDS
fisher scientific company
SS8174
Poly Bottle 4L Each for $199.38
Case of 4 Each for $797.52

SS817-1

  SDS
fisher scientific company
SS8171
Poly Bottle 1L Each for $130.38
Case of 6 Each for $782.28

SS817-20

  SDS
fisher scientific company
SS81720
PolyPac™ 20L Each for $540.00

SS817-500

  SDS
fisher scientific company
SS817500
Poly Bottle 500mL Each for $101.77
Case of 6 Each for $610.62
Description & Specifications

Specifications

Boiling Point 100°C
Melting Point 0°C
Quantity 4L
Physical Form Liquid
Color Colorless
NIST Traceable to NIST.
Vapor Pressure 14mmHg
CAS 7732-18-5
Identification Pass Test
Health Hazard 2 CAUTION!
MDL Number MFCD00003499
Health Hazard 1 Signal Word:CAUTION!
Concentration 0.1995 to 0.2005
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED POTASSIUM DICHROMATE
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0
Chemical Name or Material Sodium Thiosulfate Solution
ChemAlert Storage Symbol Gray
Name Note 0.2N
CAS 10102-17-7
Safety and Handling
 

Signal Word:CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0

CAUTION!