missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Sodium Thiosulfate Pentahydrate (Crystalline/USP/FCC/EP/BP), Fisher Chemical™
Manufacturer: Fisher Chemical S47512
Specifications
Sodium Thiosulfate Pentahydrate | |
H10Na2O8S2 | |
Pass Test (BP) | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
61475 | |
248.19 | |
99.0 to 100.5% (FCC) | |
Pass Test (USP) | |
0.002% max. (USP) | |
10ppm max. (FCC) | |
Meets Requirements (BP) | |
0.2% max. (BP) | |
32.0 to 37.0% (USP) | |
48.5°C | |
12kg |
10102-17-7 | |
MFCD00003499 | |
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
248.172 | |
CHEBI:32150 | |
Solid | |
BP/EP/FCC/USP | |
10ppm max. (BP) | |
Pass Test (BP) | |
Poly Pail | |
0.003% max. (FCC) | |
Pass Test (BP) | |
White | |
6.0 to 8.4 (EP/BP) |
Chemical Identifiers
10102-17-7 | |
248.172 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
H10Na2O8S2 | |
MFCD00003499 | |
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m | |
CHEBI:32150 | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Safety and Handling
