missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Sodium Thiosulfate Pentahydrate (Crystalline/USP/FCC/EP/BP), Fisher Chemical™
Chemical Identifiers
CAS |
10102-17-7 |
Molecular Formula |
H10Na2O8S2 |
Molecular Weight (g/mol) |
248.172 |
MDL Number |
MFCD00003499 |
InChI Key |
PODWXQQNRWNDGD-UHFFFAOYSA-L |
Synonym |
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m |
PubChem CID |
61475 |
ChEBI |
CHEBI:32150 |
IUPAC Name |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
SMILES |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
View More Specs
Products
2
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
S475212
SDS
View Documents
|
Fisher Chemical
S475212
|
2.5kg |
Poly Bottle |
Each for $249.63
|
|
Add to cart
|
|
|
S47512
SDS
View Documents
|
Fisher Chemical
S47512
|
12kg |
Poly Pail |
Each for $834.69
|
|
Add to cart
|
|
|
Description
Specifications
Chemical Identifiers
|
10102-17-7 |
|
248.172 |
|
PODWXQQNRWNDGD-UHFFFAOYSA-L |
|
61475 |
|
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
|
H10Na2O8S2 |
|
MFCD00003499 |
|
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m |
|
CHEBI:32150 |
|
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
Specifications
|
10102-17-7 |
|
MFCD00003499 |
|
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m |
|
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
|
248.172 |
|
CHEBI:32150 |
|
Solid |
|
BP/EP/FCC/USP |
|
10ppm max. (BP) |
|
Pass Test (BP) |
|
Poly Bottle |
|
0.003% max. (FCC) |
|
Pass Test (BP) |
|
White |
|
6.0 to 8.4 |
|
Sodium Thiosulfate Pentahydrate |
|
H10Na2O8S2 |
|
Pass Test (BP) |
|
PODWXQQNRWNDGD-UHFFFAOYSA-L |
|
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
|
61475 |
|
248.19 |
|
99.0 to 100.5% (FCC) |
|
Pass Test (USP) |
|
0.002% max. (USP) |
|
10ppm max. (FCC) |
|
Meets Requirements (BP) |
|
0.2% max. (BP) |
|
32.0 to 37.0% (USP) |
|
48.5°C |
|
2.5kg |
Safety and Handling
SDS