missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Thiosulfate Pentahydrate (Crystalline/USP/FCC/EP/BP), Fisher Chemical™

Manufacturer:  Fisher Chemical S475212

 View more versions of this product

Catalog No. S475212



Description

Description

Specifications

Specifications

Sodium Thiosulfate Pentahydrate
H10Na2O8S2
Pass Test (BP)
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
61475
248.19
99.0 to 100.5% (FCC)
Pass Test (USP)
0.002% max. (USP)
10ppm max. (FCC)
Meets Requirements (BP)
0.2% max. (BP)
32.0 to 37.0% (USP)
48.5°C
2.5kg
10102-17-7
MFCD00003499
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
248.172
CHEBI:32150
Solid
BP/EP/FCC/USP
10ppm max. (BP)
Pass Test (BP)
Poly Bottle
0.003% max. (FCC)
Pass Test (BP)
White
6.0 to 8.4

Chemical Identifiers

10102-17-7
248.172
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
H10Na2O8S2
MFCD00003499
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m
CHEBI:32150
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

SDS Documents
Product Certifications