Please login to your online account to display your discounted pricing

Sodium Thiosulfate, Pentahydrate, ACS Grade, 99.5 to 101.0%, LabChem™

$66.11 - $66.11

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate
CAS 10102-17-7
CAS Max % 100
Name Note Pentahydrate
Purity Grade ACS Grade
Linear Formula Na2S2O3·5H2O
Formula Weight 248.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC249901

 
lab chem
LC249901
500g Each for $66.11
Description & Specifications

Specifications

Formula Weight 248.18
Chemical Name or Material Sodium Thiosulfate
Identification Passes Test
Name Note Pentahydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Assay 99.5 to 101.0%
Solubility Soluble in water
Specific Gravity 1.73g/cm3
Storage Information Room Temperature
Linear Formula Na2S2O3·5H2O
CAS 10102-17-7
Decomposition Information Sulfur compounds
Boiling Point 100°C
Color Colorless to white
Melting Point 45°C
Quantity 500g
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Density 1.73g/cm3
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.