Please login to your online account to display your discounted pricing

Sodium thiosulfate pentahydrate, 99.5+%, ACS reagent, ACROS Organics™


Quantity 1kg
Packaging Plastic bottle

Manufacturer: fisher scientific acros  424460010

 View more versions of this product

Catalog No. AC424460010

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 248.18
Physical Form Crystals
Purity Grade ACS Reagent Grade
Quantity 1kg
Color White
Assay 99.5 to 101.0%
pH 6.0 to 8.4 (5% soln. at 25°C)
Density 1.7200g/mL
Decomposition Information >45°C
Merck Index 15, 8821
Linear Formula Na2S2O3·5H2O
MDL Number MFCD00149186
Chemical Name or Material Sodium thiosulfate pentahydrate
Molecular Formula Na2O3S2·5H2O
CAS 10102-17-7
Nitrogen Compounds 0.002% max.
Sulfide Passes Test (limit about 1ppm)
Packaging Plastic bottle
Insoluble Matter 0.005% max.
Sulfate and Sulfite 0.1% max.
Assay Percent Range 99.5+%

Specifications

Chemical Name or Material Sodium thiosulfate pentahydrate
CAS 10102-17-7
Purity Grade ACS Reagent Grade
Assay 99.5 to 101.0%
Assay Percent Range 99.5+%
Molecular Formula Na2O3S2·5H2O
Linear Formula Na2S2O3·5H2O
Formula Weight 248.18
MDL Number MFCD00149186