Please login to your online account to display your discounted pricing

Sodium Thiosulfate Pentahydrate, 99.5-101.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals


Quantity 1kg

Manufacturer: mp biomedicals  19144701

 View more versions of this product

Catalog No. ICN19144701

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 248.17
Quantity 1kg
Physical Form Crystalline
CAS 10102-17-7
Chemical Name or Material Sodium Thiosulfate Pentahydrate
Molecular Formula Na2S2O3 · 5H2O
Assay Percent Range 99.5-101.0%
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Sodium Thiosulfate Pentahydrate
CAS 10102-17-7
Purity Grade ACS Reagent Grade
Physical Form Crystalline
Assay Percent Range 99.5-101.0%
Molecular Formula Na2S2O3 · 5H2O
Formula Weight 248.17
Accessories

Room Temperature