Please login to your online account to display your discounted pricing

Sodium Thiosulfate Pentahydrate, 99.5-101.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals

$127.63 - $351.62

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate Pentahydrate
CAS 10102-17-7
Purity Grade ACS Reagent Grade
Physical Form Crystalline
Assay Percent Range 99.5-101.0%
Molecular Formula Na2S2O3 · 5H2O
Formula Weight 248.17
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19144701

 
mp biomedicals
19144701
1kg Each for $127.63

ICN19144705

 
mp biomedicals
19144705
5kg Each for $351.62
Description & Specifications

Specifications

Purity Grade ACS Reagent Grade
Formula Weight 248.17
Quantity 1kg
Physical Form Crystalline
Chemical Name or Material Sodium Thiosulfate Pentahydrate
Assay Percent Range 99.5-101.0%
Molecular Formula Na2S2O3 · 5H2O
CAS 10102-17-7
Recommended Storage Room Temperature
Safety and Handling

Room Temperature