missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium thiosulfate pentahydrate, 99.5-101.0%, ACS, Alfa Aesar™

Maximum level of impurities: Insoluble matter 0.005%, pH of a 5% solution 6.0-8.4 at 25°, Nitrogen compounds (as N) 0.002%, Sulfate and sulfite (as SO4) 0.1%, Sulfide P.T. (limit about 1ppm)

Manufacturer:  Alfa Aesar 14518A3

 View more versions of this product

Catalog No. AA14518A3



Specifications

Specifications

Sodium thiosulfate pentahydrate
10102-17-7
H10Na2O8S2
Colorless crystalline
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
61475
248.17 (158.10 Anhydrous)
ACS
Colorless
For ACS analysis
99.5-101.0%
MFCD00149186
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
248.172
CHEBI:32150
Crystalline
Odorless
2kg

Chemical Identifiers

10102-17-7
248.172
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
H10Na2O8S2
MFCD00149186
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

EINECSNumber : 231-867-5

Documents