missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Thiosulfate, For Iodine Absorption Number, Certified, 0.0394N ±0.0003N (0.0394M), LabChem™

$96.46 - $96.46

Chemical Identifiers

CAS 7732-18-5,10102-17-7,497-19-8
CAS Max % 99,0.98,0.02
Linear Formula Na2S2O3·5H2O
Formula Weight 248.18
Chemical Name or Material Sodium Thiosulfate, For Iodine Absorption Number
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC250404 Lab Chem
LC250404
4L Each for $96.46
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,10102-17-7,497-19-8
Na2S2O3·5H2O
Sodium Thiosulfate, For Iodine Absorption Number
99,0.98,0.02
248.18
Certified Grade

Specifications

248.18
Clear, colorless
Passes Test
7732-18-5,10102-17-7,497-19-8
Poly Bottle
0.0394N ±0.0003N (0.0394M)
Sulfur compounds; Carbon dioxide
Sodium Thiosulfate, For Iodine Absorption Number
0.02,0.98,99
4L
Soluble in water
Liquid
Room Temperature
Na2S2O3·5H2O
Traceable to NIST
Certified Grade

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.