missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Thiosulfate (Crystalline/USP/FCC), Fisher Chemical™

Manufacturer:  Fisher Chemical FLS474500

 View more versions of this product

Catalog No. S474-500




Description

Description

Applications include:Dechlorinination and/or debromination of waterPhotographic processingBacteriological water assessmentAntidote to cyanide poisoningMeasuring the volume of extracellular body fluid and the renal glomerular filtration rateTreatment of calciphylaxis in hemodialysis patients with end-stage kidney disease

Sodium thiosulfate has been studied in many applications including dechlorinination and or debromination of water, and photographic processing. Because of its ability to remove chlorine, it is also used in bacteriological water assessment, as it promotes the survival of coliform organisms by neutralizing residual chlorine. It is also used as an antidote to cyanide poisoning, and in measuring the volume of extracellular body fluid and the renal glomerular filtration rate. It has been studied as treatment of calciphylaxis in hemodialysis patients with end-stage kidney disease.
Specifications

Specifications

Sodium Thiosulfate
White
6.0 to 8.4
H10Na2O8S2
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
248.172
CHEBI:32150
Solid
FCC/USP
2mg/kg max. (FCC)
Meets Requirements (USP)
32.0 to 37.0% (USP)
>45°C
10102-17-7
48.5°C
500g
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
61475
248.19
99.0 to 100.5% (FCC)
Pass Test (FCC)
Poly Bottle
0.003% max. (FCC)
Pass Test (USP)

Chemical Identifiers

10102-17-7
248.172
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
H10Na2O8S2
MFCD00149186
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m
CHEBI:32150
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:1

SDS Documents
Product Certifications
Promotions

Promotions