Please login to your online account to display your discounted pricing

Sodium Thiosulfate, Certified, 1.00N ±0.01N (1.0M), LabChem™

$31.49 - $391.69

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate
CAS 7732-18-5
CAS Max % 75.16
CAS 10102-17-7
CAS Max % 24.82
CAS 497-19-8
CAS Max % 0.02
Purity Grade Certified Grade
Linear Formula Na2S2O3·5H2O
Formula Weight 248.18
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC251001

 
lab chem
LC251001
500mL Each for $31.49

LC251002

 
lab chem
LC251002
1L Each for $44.03

LC251004

 
lab chem
LC251004
4L Each for $128.69

LC251005

 
lab chem
LC251005
20L Each for $391.69
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 248.18
Chemical Name or Material Sodium Thiosulfate
CAS Max % 0.02
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula Na2S2O3·5H2O
CAS 7732-18-5
Decomposition Information Sulfur compounds; Carbon dioxide
Color Clear, colorless
Quantity 500mL
CAS Max % 75.16
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 10102-17-7
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 24.82
Concentration 1.00N ±0.01N (1.0M)
Traceability to NIST Traceable to NIST
CAS 497-19-8
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.