missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Thiosulfate, Certified, 1.00N ±0.01N (1.0M), LabChem™
Supplier: LabChem LC251004
Specifications
Sodium Thiosulfate | |
Colorless | |
Liquid | |
H10Na2O8S2 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium pentahydrate sulfanidesulfonate | |
61475 | |
248.18 | |
1.00N ±0.01N (1.0M) | |
Poly Bottle |
10102-17-7,7732-18-5,497-19-8 | |
24.82,75.16,0.02 | |
4 L | |
Na2S2O3·5H2O | |
Soluble in water | |
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O | |
248.17 | |
CHEBI:32150 | |
Certified | |
Passes Test | |
Sulfur compounds; Carbon dioxide |
Chemical Identifiers
10102-17-7 | |
248.17 | |
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
CHEBI:32150 | |
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O |
H10Na2O8S2 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium pentahydrate sulfanidesulfonate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature