missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Thiosulfate, Certified, 0.200N ±0.002N (0.2M), LabChem™

Manufacturer:  LabChem LC250651

 View more versions of this product

Catalog No. LC250651



Specifications

Specifications

Sodium Thiosulfate
95.02,4.96,0.02
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Liquid
0.200N ±0.002N (0.2M)
Poly Bottle
Room Temperature
Colorless
7732-18-5,10102-17-7,497-19-8
Na2S2O3·5H2O
248.18
Certified
Passes Test
Soluble in water
Sulfur compounds; Carbon dioxide
500mL

Chemical Identifiers

7732-18-5
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Documents
Promotions

Promotions