Please login to your online account to display your discounted pricing

Sodium Thiosulfate, Anhydrous, Granular, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  395305

 View more versions of this product

Catalog No. 02004262

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 158.11
Chemical Name or Material Sodium Thiosulfate
CAS 7772-98-7
Name Note Anhydrous
Packaging Wide Mouth Amber Glass Bottle
Purity Grade BAKER ANALYZED™ Reagent
Physical Form Granular
Specific Gravity 1L = 1.67kg
Molecular Formula Na2S2O3
Quantity 2.5kg

Specifications

Chemical Name or Material Sodium Thiosulfate
CAS 7772-98-7
Name Note Anhydrous
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Na2S2O3
Formula Weight 158.11