Please login to your online account to display your discounted pricing

Sodium Thiosulfate, Anhydrous, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: fisher scientific acros  202870025

 View more versions of this product

Catalog No. AC202870025

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium thiosulfate
CAS 7772-98-7
Purity Grade Extra Pure
Physical Form Crystalline Powder
Assay ≥98.0% (on dry substance)
Assay Percent Range 98.5%
Molecular Formula Na2O3S2
Linear Formula Na2S2O3
Formula Weight 158.1
Chlorine (Cl) 1500ppm max.
Heavy Metals (as Pb) 50ppm max.
Iron (Fe) 50ppm max.
Loss on Drying 2% (105°C, 2 hrs) max.
Packaging Plastic bottle
Sulfate and Sulfite 1% max.
Color White
pH 6.0 to 8.5 (5% soln. at 20°C)
Quantity 2.5kg

Specifications

Chemical Name or Material Sodium thiosulfate
CAS 7772-98-7
Purity Grade Extra Pure
Assay ≥98.0% (on dry substance)
Assay Percent Range 98.5%
Molecular Formula Na2O3S2
Linear Formula Na2S2O3
Formula Weight 158.1