Please login to your online account to display your discounted pricing

Sodium Thiosulfate, Anhydrous, 97% min., EMD Millipore


Quantity 500g

Manufacturer: emd millipore corporation  SX08201

 View more versions of this product

Catalog No. MSX08201

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 158.11
Quantity 500g
Color White
Assay Percent Range 97%, min.
Linear Formula Na2S2O3
Synonym Sodium Hyposulfate, pentahydrate
Chemical Name or Material Sodium Thiosulfate, Anhydrous
Molecular Formula Na2S2O3
CAS 7772-98-7
Physical Form Powder

Specifications

Chemical Name or Material Sodium Thiosulfate, Anhydrous
CAS 7772-98-7
Physical Form Powder
Assay Percent Range 97%, min.
Molecular Formula Na2S2O3
Linear Formula Na2S2O3
Formula Weight 158.11
Synonym Sodium Hyposulfate, pentahydrate