missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Thiosulfate, Anhydrous, 97% min., EMD Millipore

$127.71 - $1,119.54

Chemical Identifiers

CAS 7772-98-7
Physical Form Powder
Assay Percent Range 97%, min.
Molecular Formula Na2S2O3
Linear Formula Na2S2O3
Formula Weight 158.11
Synonym Sodium Hyposulfate, pentahydrate
Chemical Name or Material Sodium Thiosulfate, Anhydrous
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MSX08201 Emd Millipore Corporation
SX08201
500g Each for $127.71
MSX08205 Emd Millipore Corporation
SX08205
12kg Each for $1,119.54
Specifications Description & Specifications

Chemical Identifiers

7772-98-7
97%, min.
Na2S2O3
Sodium Hyposulfate, pentahydrate
Powder
Na2S2O3
158.11
Sodium Thiosulfate, Anhydrous

Specifications

158.11
White
Na2S2O3
Na2S2O3
Powder
500g
97%, min.
Sodium Hyposulfate, pentahydrate
7772-98-7
Sodium Thiosulfate, Anhydrous