Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 8% (w/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate
CAS 7732-18-5
CAS Min % 91.0
CAS Max % 94.0
CAS 10102-17-7
CAS Min % 7.26
CAS Max % 7.56
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R78672001A

 
ricca chemical company
RIR78672001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R78672004A

 
ricca chemical company
RIR78672004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 1L
CAS 7732-18-5
CAS Min % 91.0
CAS 10102-17-7
Packaging Poly natural
CAS Min % 7.26
CAS Max % 7.56
Chemical Name or Material Sodium Thiosulfate
Concentration or Composition Notes 8% (w/v)
Solvent or Matrix Aqueous Solution
CAS Max % 94.0