Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$127.28 - $225.81

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate
CAS 7772-98-7
Name Note 1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula Na2S2O3
Formula Weight 158.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004257

 
avantor performance
565402
Narrow Mouth Polyethylene Bottle 1L Each for $127.28

02004258

 
avantor performance
565403
Cubetainer 4L Each for $225.81
Description & Specifications

Specifications

Formula Weight 158.11
Chemical Name or Material Sodium Thiosulfate
CAS 7772-98-7
Name Note 1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Quantity 1L
Specific Gravity 1L = 1.12kg
Molecular Formula Na2S2O3