Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 0.28 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 10102-17-7
CAS Min % 4.79
CAS Max % 4.98
CAS 497-19-8
CAS Min % 0.04
CAS Max % 0.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

79621

 
ricca chemical company
RI79621
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

796232

 
ricca chemical company
RI796232
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Sodium Thiosulfate
CAS Min % 93.0
Concentration or Composition Notes 0.28Normal
Packaging Poly natural
CAS Max % 0.04
CAS Max % 97.0
CAS 497-19-8
CAS Min % 0.04
CAS Max % 4.98
CAS Min % 4.79
CAS 7732-18-5
CAS 10102-17-7