missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Thiosulfate, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 563702
Specifications
Sodium Thiosulfate | |
7772-98-7 | |
Na2O3S2 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
158.10 | |
158.11 | |
Narrow Mouth Polyethylene Bottle |
0.1N Volumetric Solution | |
1 L | |
MFCD00003499 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
disodium sulfanidesulfonate | |
24477 | |
BAKER ANALYZED™ Reagent | |
1L = 1.01kg |
Chemical Identifiers
7772-98-7 | |
158.10 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
24477 | |
[Na+].[Na+].[O-]S([S-])(=O)=O |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
disodium sulfanidesulfonate |