missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 0.1N (N/10), Ricca Chemical

$103.36 - $468.58

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7950-1
SDS View Documents
Ricca Chemical Company
RI79501
4L Poly natural
Each for $103.36
 
79-503-2CS
SDS View Documents
Ricca Chemical Company
RIR795000012X1A
12L Poly natural
Each for $468.58
 
79-501-CS
SDS View Documents
Ricca Chemical Company
RIR79500004X4A
16L Poly natural
Each for $354.33
 
Description

Description

Specifications

Chemical Identifiers

10102-17-7
248.172
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m
CHEBI:32150
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate

Specifications

10102-17-7,497-19-8,7732-18-5
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
61475
Odorless to slight solvent odor
Solubility in Water: Infinite
4L
0.1000 ± 0.0001 N at 20°C
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
248.172
CHEBI:32150
Poly natural
Aqueous Solution
Sodium Thiosulfate, 0.1N (N/10)
SDS