missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Thiosulfate, 0.1N (N/10), Ricca Chemical
Manufacturer: Ricca Chemical Company RIR79500004X4A
Specifications
Sodium Thiosulfate | |
10102-17-7,497-19-8,7732-18-5 | |
H10Na2O8S2 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
61475 | |
Odorless to slight solvent odor | |
Solubility in Water: Infinite | |
16L |
0.1N (N/10) | |
0.1000 ± 0.0001 N at 20°C | |
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
248.172 | |
CHEBI:32150 | |
Poly natural | |
Aqueous Solution |
Chemical Identifiers
10102-17-7 | |
248.172 | |
ametox, antichlor, ccris 3952, disodium thiosulfate pentahydrate, sodium hyposulfite pentahydrate, sodium thiosulfate pentahydrate, sodium thiosulfate, pentahydrate, thiosulfuric acid, disodium salt, pentahydrate, tinver, unii-hx1032v43m | |
CHEBI:32150 | |
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
H10Na2O8S2 | |
PODWXQQNRWNDGD-UHFFFAOYSA-L | |
61475 | |
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |