Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 0.1 N N/10) Standard Volumetric Solution, Macron Fine Chemicals™

macron

$228.70 - $228.70

Chemical Identifiers

CAS 7772-98-7
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MK46821

 
avantor performance
468260
1L Each for $228.70
Description & Specifications

Specifications

CAS 7772-98-7
Physical Form Liquid
Quantity 1L
Appearance Clear, colorless solution
Normality 0.0995 to 0.1005 N