Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 0.1 N N/10) Standard Volumetric Solution, Macron Fine Chemicals™


macron

Quantity 20L

Manufacturer: avantor performance  468205

 View more versions of this product

Catalog No. MK468220

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

CAS 7772-98-7
Physical Form Liquid
Quantity 20L
Appearance Clear, colorless solution

Specifications

CAS 7772-98-7
Physical Form Liquid