Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 0.0200 Normal (N/50), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate
Name Note 0.0200 Normal (N/50)
CAS 10102-17-7
CAS 497-19-8
CAS 7732-18-5
Physical Form Liquid
Assay Percent Range 0.02000 ± 0.00001 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R78950004C

 
ricca chemical company
RIR78950004C
Amber glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R78950004F

 
ricca chemical company
RIR78950004F
Cubitainer 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 4L
Solvent or Matrix Aqueous Solution
Chemical Name or Material Sodium Thiosulfate
Name Note 0.0200 Normal (N/50)
Assay Percent Range 0.02000 ± 0.00001 N at 20°C
CAS 10102-17-7
CAS 497-19-8
Odor Odorless to slight solvent odor
Packaging Amber glass
Solubility Solubility in Water: Infinite
CAS 7732-18-5