missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium thiosulfate, 0.01N, Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance H36207

 View more versions of this product

Catalog No. MKH3624

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium thiosulfate
Na2O3S2
AKHNMLFCWUSKQB-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
24477
Crystals
White
7772-98-7
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
[O-]S(=O)(=S)[O-].[Na+].[Na+]
158.097
248.18
poly bottle
4L

Chemical Identifiers

7772-98-7
158.097
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Na2O3S2
AKHNMLFCWUSKQB-UHFFFAOYSA-L
24477
[O-]S(=O)(=S)[O-].[Na+].[Na+]
Documents