Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 0.01N, LaMotte™

$241.69 - $241.69

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate
CAS 10102-17-7
CAS 497-19-8
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

03-920-376

 
lamotte chemical prod co
3819N
3800mL Each for $241.69
Description & Specifications

Specifications

Quantity 3800mL
Analyte or Component Names Sodium thiosulfate,Sodium carbonate
Solvent or Matrix Water
Physical State Liquid
Color Pale yellow
CAS 10102-17-7
CAS 497-19-8
Chemical Name or Material Sodium Thiosulfate