Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 0.0143 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Thiosulfate
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 10102-17-7
CAS Min % 0.28
CAS Max % 0.3
CAS 497-19-8
CAS Min % 0.0023
CAS Max % 0.0023
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7892-5

 
ricca chemical company
RI78925
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R78920004A

 
ricca chemical company
RIR78920004A
Natural poly 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R78920001A

 
ricca chemical company
RIR78920001A
Natural poly 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 20L
CAS Min % 97.0
CAS Max % 0.3
Chemical Name or Material Sodium Thiosulfate
CAS 7732-18-5
CAS Max % 100.0
Concentration or Composition Notes 0.0143Normal
CAS Min % 0.28
CAS 497-19-8
CAS Min % 0.0023
CAS 10102-17-7
CAS Max % 0.0023
Packaging Cubitainer