Please login to your online account to display your discounted pricing

Sodium Thiosulfate, 0.0143 Normal, Ricca Chemical


Quantity 1L
Packaging Natural poly

Manufacturer: ricca chemical company  RIR78920001A

 View more versions of this product

Catalog No. R78920001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 1L
CAS 7732-18-5
Concentration or Composition Notes 0.0143Normal
CAS 497-19-8
CAS Max % 0.3
CAS Max % 0.0023
CAS Min % 0.0023
CAS Max % 100.0
CAS Min % 97.0
CAS Min % 0.28
CAS 10102-17-7
Packaging Natural poly
Chemical Name or Material Sodium Thiosulfate

Specifications

Chemical Name or Material Sodium Thiosulfate
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 10102-17-7
CAS Min % 0.28
CAS Max % 0.3
CAS 497-19-8
CAS Min % 0.0023
CAS Max % 0.0023
Physical Form Liquid